Specifications
- Size:100 mg
- Protein/Peptide Name:Aprotinin
- Cat. No.:82022-378
- Supplier no.:786-046
Specifications
About this item
A selection of individual protease inhibitors are offered for researchers who wish to design their own cocktails, supplement existing cocktails, or screen for specific proteases
- Inhibits serine proteases (plasmin, kalikrein, trypsin and chymotrypsin).
Also available is the Protease Inhibitor Set, which contains all 12 of our individual protease inhibitors, each in a ready-to-use solution at a 100X concentration. The 1X concentration of the protease inhibitors is designed to give >90% inhibition in crude tissue extracts. Various concentrations and/or combinations of protease inhibitors may be used to inhibit a broad spectrum of protease activity.
G-Biosciences offers the following individual protease inhibitors as a dry powder: AEBSF, ALLN, Antipain dihydrochloride, Aprotinin, Bestatin, Chymostatin, E-64, EDTA-Na2, Leupeptin, Pepstatin, Phosphoramidon, and PMSF.