
Paramat Extra, Solidification point: 56-58°C embedding agent for histology
Supplier: Electron Microscopy Sciences
Certificates
About this item
CAS Number:8002-74-2
:8012-95-1
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:Milky-white, pastilles
- Crystalline Structure under the microscope:Needles & Platelets
- Effect of heating (@60°C/100g):Clear melt within max. 16 hrs
- Solubility in Xylene (@20°C/1g in 50 ml):Clear solution within max. 16 hrs
- Cutting results for 4*m sections:Over 75% usable single; over 65% usable serial
- Solubility in Xylene (section):Max. 45 sec
- Melting Point:56-58°C
- Solidification Point:58-56°C
- Tears and Vascuoles:None
- Adhesion of sections Very good; protein, to microscope slides:glycerol etc. is not required to hold the section in place
- Free acid content:Max. 0.01%
- Block Structure:No mottling, no shrinking
- Block Solidification (room Temperature):About 2 hrs
- DMSO content:Less than 0.1%