Fluorescent Dye 488-I Phalloidin, Abnova
Catalog # 10678-000
Supplier: Abnova
CAS Number:
Specifications
- Description:Fluorescent Dye 488-I Phalloidin
- Size:300 Reactions
- Storage Temperature:–20 °C
- Application:Phalloidin conjugate
- Ex/em:491/514 nm
- Cat. No.:10678-000
- Supplier no.:U0281
Specifications
About this item
Fluorescence Dye 488-I conjugates are significantly brighter than fluorescein conjugates and much more photostable.
- Spectral Equivalent: Alexa Fluor 488, Cy2, DyLight 488, FITC
- Molar Extinction Coefficient: ≥70,000 M-1cm-1
The fluorescence of Fluorescence Dye 488-I is not affected by pH between 4 - 10. Compared to Alexa Fluor 488, Fluorescence Dye 488-I of single isomer has much hihger purity, bringing better performance consistancy from batch to batch.