Human, Mouse, Rat, Non-Human Primate Desoxyribonuclease I (DNase I)
Deoxyribonuclease (DNase) I solution is a cell dissociation reagent useful to reduce or prevent the clumping of concentrated or cryopreserved cell suspensions following thawing.
Contains: Bovine pancreatic deoxyribonuclease I (1 mg/mL) and phosphate-buffered saline
Subtype: Enzymatic
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
- Species:HumanMouseNon-human primateRat
- Enzyme Name:DNase I Solution (1 mg/ml)
- Enzyme Synonyms:DNA endonuclease; DNA nuclease; Deoxyribonucleic phosphatase; Pancreatic DNase; Thymonuclease
Frequently Bought Together