About this item
pluriStrainer® is a sterile sieving device to obtain real single cell suspensions or to remove cell aggregates.
- Unique design features improved ventilation to avoid clogging
- Physical dissociation of primary tissue
- Faster and easier alternative to gauze filtration
- Short time incubation of cells/cytokine
- Sieving of complex and viscous liquids
- PET mesh(Woven) material
Obtaining of real single cell suspension after enzymatic digestion of mammary tissue and organoids.
Preparation of single cell suspension of blood cells from bone marrow, pancreas, thymus, lymph nodes and others. Preparation of real single cell suspension for flow cytometry.
Fits into 50 ml conical centrifuge tube (standard).
Housing material is made of LDPE (Low Density Polyethylene).
pluriStrainer® Set 2 includes 4 × (40, 60, 70, 85, 100 and 200 µm) meshes.