Centrifuge 5910 R, Refrigerated, Eppendorf
Supplier: Avantor
Certificates
About this item
Centrifuge 5910® R combines a high capacity of up to 4×750 ml.
- Swing-bucket rotors
- Plate rotor options for centrifugation of all types of MTP, PCR or deepwell plates
- Fixed-angle rotors for high-speed molecular biology applications
- Dynamic compressor control (DCC) technology for precise cooling performance
- Refrigeration system with advanced temperature management
FastTemp pro® function allows for automatic precooling based on pre-programmable time and date.
Rotors
Product Information
Frequently Bought Together