
Cesium chloride, anhydrous ≥99.9%, Reagent Grade, free-flowing, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7647-17-8
:
Formula:CsCl
MDL Number:MFCD00010955
:168.36 g/mol
Melting Pt:642 °C
Density:3.98 g/cm³ (20 °C)
EINECS:231-600-2
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay:99 - 101 %
- Metal trace analysis (ICP):Max. 1500 ppm
Specification Test Results
Frequently Bought Together