Order Entry
Puerto Rico
ContactUsLinkComponent
Organic Reference Standards, Azithromycins, RS, USP
Organic Reference Standards, Azithromycins, RS, USP
Catalog # 101094-348
Supplier:  USP
CAS Number:  
Organic Reference Standards, Azithromycins, RS, USP
Catalog # 101094-348
Supplier:  USP
Supplier Number:  1046056
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • CAS No.:
    117772-70-0
  • Description:
    Azithromycin
  • Size:
    100 mg
  • Formula:
    C₃₈H₇₂N₂O₁₂.₂H₂O
  • Description:
    Azithromycin dihydrate
  • Cat. No.:
    101094-348
  • Supplier no.:
    1046056

Specifications

About this item