Order Entry
Puerto Rico
ContactUsLinkComponent
miRNA QRT-PCR Detection Kit, Agilent Technologies
miRNA QRT-PCR Detection Kit, Agilent Technologies
Catalog # 97066-464
CAS Number:  
undefined
miRNA QRT-PCR Detection Kit, Agilent Technologies
Catalog # 97066-464
Supplier Number:  600584
CAS Number:  

Specifications

  • Application:
    RT-qPCR dye
  • Description:
    miRNA qRT-PCR master mix detection kit
  • Size:
    200 reactions
  • Cat. No.:
    97066-464
  • Supplier no.:
    600584
  • Details:
    Kit provides reagents for the synthesis of first-strand cDNA from the tailed miRNAs as well as a qPCR master mix containing all the necessary enzymes, dyes, and reagents, to give the utmost specificity for miRNA qPCR

Specifications

About this item

Polyadenylation of miRNA from total RNA and synthesis of first-strand cDNA from tailed miRNAs.

  • Fast and accurate detection of mature miRNA
  • Differentiate miRNA that differ by just a single nucleotide for enhanced sensitivity
  • Sensitive detection down to 10 copies is enabled
  • Rapid scanning performance is ensured, with highly accurate results in as little as three hours
  • Includes all necessary miRNA Specific Forward Primers

Agilent miRNA qPCR Detection Reagents are high-specificity reagents for the polyadenylation of microRNAs (miRNAs) from total RNA. The miRNA qPCR Detection Reagents also enable the synthesis of first-strand cDNA from the tailed miRNAs.

The reagent kit includes miRNA Specific Forward Primers and all necessary microRNA qPCR MM Detection Reagents for accurate analysis. Highly sensitive detection down to ten copies is enabled, and miRNA that differs by even a single nucleotide can be identified.