Specifications
- Refine target molecule:Viral DNA
- Target molecule:DNA
- Description:AdEasy Virus Purification Kit
- Size:1 Kit
- Cat. No.:99901-386
- Supplier no.:240243
Specifications
About this item
The AdEasy Virus Purification Kit allows the purification and concentration of Adenovirus (from Ad5 strains) with Sartobind® syringe filters containing an ion exchange membrane adsorber that selectively binds adenoviral particles. Once bound, viral particles can be further purified by washing away nonspecifically-bound proteins before elution. Concentrated and purified viral particles can be obtained in 2 to 3 hours, in contrast to traditional CsCl gradient centrifugation which typically takes 12 to 48 hours.
- Rapid alternative to cesium chloride (CsCl) density gradient methods
- No cesium chloride or ultracentrifugation necessary
- Increased infection rates and cell viability
AdEasy virus purification kits rapid membrane chromatography is based method for the purification and concentration of type 5 adenoviral stocks.