Sodium molybdate(VI) dihydrate 99-100.5% (dried basis), Puriss., meets analytical specification of Ph.Eur., Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:10102-40-6
:
Formula:Na₂MoO₄·2H₂O
MDL Number:MFCD00149170
:241.95 g/mol
Melting Pt:100 °C (–H₂O of crystall.)
Density:2.700 g/cm³ (20 °C)
Storage Temperature:Ambient
Specification Test Results
- Assay (calculated to the dried substance):99.0 - 100.5 %
- Loss on drying (140°C, 3h):14.0 - 16.0 %
- Ammonium (NH₄):Max. 10 ppm
- Iron (Fe):Max. 0.001 %
- Heavy metals (as Pb):Max. 10 ppm
- Chloride (Cl):Max. 50 ppm
- Phosphate (PO₄):Max. 200 ppm
- Sulfate (SO₄):Max. 0.05 %
- Appearance of the solution:complying
- Residual solvents:complying