Order Entry
Puerto Rico
ContactUsLinkComponent
Tissue-Tek® Prisma® Plus Automated Slide Stainer, Sakura Finetek
Tissue-Tek® Prisma® Plus Automated Slide Stainer, Sakura Finetek
Catalog # 76176-846
Supplier:  Sakura Finetek
undefined
Tissue-Tek® Prisma® Plus Automated Slide Stainer, Sakura Finetek
Catalog # 76176-846
Supplier:  Sakura Finetek
Supplier Number:  6170

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Sakura Prisma® Plus Automated Slide Stainer
  • Dimensions:
    49.2W×28D×25H"
  • Cat. No.:
    76176-846
  • Electrical:
    115 VAC, 50/60 Hz, 9 A
  • Application:
    Haemotoxylin and Eosin, Papanicolaou (PAP), Non-GYN

Specifications

About this item

High throughput slide stainer, capable of processing up to 530 slides per hour. Continuous loading of slides, with intuitive software to run multiple staining protocols simultaneously.

  • Capable of up to 50 user-defined staining protocols
  • Capable of prioritizing stat run over standard
  • Color touchscreen with choice of 50 colors for screen stain assignments
  • Barcode reader to track reagent usage and alerts for expired reagent
  • iSupport service link monitors service issues

Standard solution configuration means 30 reagent positions and 60 slides per run or expanded configuration is 52 positions and 20 slides per run.

Connect the Prisma® Plus with the Tissue-Tek® Film® or G2 coverslippers for a fully automated, integrated, walkaway staining process from slide baking to slide drying.

Certifications: IEC 61010-1 2nd ed., CAN/CSA C22.2 No. 61010-1, UL 61010-1.
Delivery: Includes with operating manual, twenty-two 820 mL solution reservoirs, ten 20-slide baskets and handles, four wash reservoirs, two fume filters, compact flash card and reagent lids.