Order Entry
Puerto Rico
ContactUsLinkComponent
 
EDTA disodium salt 0,0178 mol/l (N/28; 1 ml = 1°dH/100 ml H₂O) in aqueous solution volumetric solution, for hardness of water determinations, Fluka™
Catalog # BJ-34543-1L
CAS Number:  
EDTA disodium salt 0,0178 mol/l (N/28; 1 ml = 1°dH/100 ml H₂O) in aqueous solution volumetric solution, for hardness of water determinations, Fluka™
Catalog # BJ-34543-1L
Supplier Number:  34543-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 L
  • Cat. No.:
    BJ-34543-1L

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001

Specification Test Results

About this item

 :
 :336.21 g/mol
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672