EDTA disodium salt 0.2 M EDTA-Na2 volumetric solution, for complexometry, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:139-33-3
:
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672
:336.21 g/mol
Storage Temperature:Ambient
EINECS:382-266-9
Specification Test Results
- Factor:0.999 - 1.001