ContactUsLinkComponent
Trubore Pressure Bearing with Ace-Threds, 19 mm, Ace Glass
Trubore Pressure Bearing with Ace-Threds, 19 mm, Ace Glass
Catalog # 76214-466
Supplier:  Ace Glass
Small Business Enterprise
undefined
Trubore Pressure Bearing with Ace-Threds, 19 mm, Ace Glass
Catalog # 76214-466
Supplier:  Ace Glass
Supplier Number:  8049-02
Small Business Enterprise
Specifications
  • Description:
    Trubore Pressure Bearing, #25 – #25 PTFE Coupling, #25 – #25 Glass Bearing
  • Cat. No.:
    76214-466
  • Shaft length:
    19 mm stir shaft
Specifications
About this item
Glass bearing designed for use with Ace-Thred pressure reactors and ACE polished glass 19 mm stir shafts.
  • Bearing made of high-quality borosilicate glass
  • Both the top bushing and bottom coupling are PTFE
  • Maximum pressure is 30 psig at ambient temperature
  • Maximum rpm: 350