ContactUsLinkComponent
Laboratory Flask Mixer, Yamato
Laboratory Flask Mixer, Yamato
Supplier:  Yamato Scientific
Certificates
undefined
Laboratory Flask Mixer, Yamato
Supplier:  Yamato Scientific
Specifications
  • Catalog No:
  • 76209-608
  • 76209-708
  • Max. Stirring Capacity H₂O:
  • 50 - 5000 ml
  • 50 - 5000 ml
  • Speed Range:
  • 50 - 1000 rpm
  • 50 - 1000 rpm
  • Max. Torque:
  • 10 Ncm
  • 10 Ncm
  • Permissible Ambient Temperature:
  • 5…35 °C
  • 5…35 °C
Specifications