ContactUsLinkComponent
Programmable Refrigerant Incubators, Yamato
undefined
Programmable Refrigerant Incubators, Yamato
  • Catalog No:
  • 76209-634
  • 76209-632
  • 76209-630
  • 76209-628
  • 76209-626
  • 76209-624
  • 76209-622
  • 76209-620
  • 76209-618
  • 76209-636
  • Temperature Setting Accuracy:
  • ±0.3 °C (refrigerator in continuous operation)
    ±1.0 °C (refrigerator in cycle operation)
  • ±0.3 °C (refrigerator in continuous operation)
    ±1.0 °C (refrigerator in cycle operation)
  • ±0.3 °C (refrigerator in continuous operation)
    ±1.0 °C (refrigerator in cycle operation)
  • ±0.3 °C (refrigerator in continuous operation)
    ±1.0 °C (refrigerator in cycle operation)
  • ±0.3 °C (refrigerator in continuous operation)
    ±1.0 °C (refrigerator in cycle operation)
  • ±0.3 °C (refrigerator in continuous operation)
    ±1.0 °C (refrigerator in cycle operation)
  • ±0.3 °C (refrigerator in continuous operation)
    ±1.0 °C (refrigerator in cycle operation)
  • ±0.3 °C (refrigerator in continuous operation)
    ±1.0 °C (refrigerator in cycle operation)
  • ±0.3 °C (refrigerator in continuous operation)
    ±1.0 °C (refrigerator in cycle operation)
  • ±0.3 °C (refrigerator in continuous operation)
    ±1.0 °C (refrigerator in cycle operation)
  • Temperature Range:
  • –10…+50 °C
  • –10…+50 °C
  • –10…+50 °C
  • –10…+50 °C
  • –10…+50 °C
  • –10…+50 °C
  • –10…+50 °C
  • –10…+50 °C
  • –10…+50 °C
  • –10…+50 °C
  • No. of shelf rest steps/pitch:
  • 13 Steps/30 mm
  • 23 Steps/30 mm
  • 23 Steps/30 mm
  • 13 Steps/30 mm
  • 13 Steps/30 mm
  • 23 Steps/30 mm
  • 23 Steps/30 mm
  • 13 Steps/30 mm
  • 13 Steps/30 mm
  • 13 Steps/30 mm