Multi-Function Immersion Thermostatic Devices, Yamato
Supplier: Avantor
Certificates
About this item
Multi-function programmable immersion constant temperature thermostatic device with propeller or water jet stirring designed for various usage.
- TRIAC zero-cross type heater circuit
- Water jet strength can be adjusted in 10 patterns
- Available clamp thickness: up to 35 mm
- No protrusions for easy setup
- Stainless pipe heater
The basic model has a fixed temperature operation. The multi-function models feature fixed temperature and program operation. Safety functions include self-diagnostic functions, a circuit protector, water level detection sensor (float type), and buzzer alarm for abnormal conditions.
Users can adjust the setting with digital keys and a green LED display indicates the temperature.
Delivery: Testing bath model BY100 (polypropylene) included.