Recombinant Leucine-rich repeats and immunoglobulin-like domains protein 1
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
- Consistent Sizing options
- Datasheet Included
- High Purity, Low Endotoxin levels
Recombinant Mouse Leucine-rich Repeats and Immunoglobulin-like Domains Protein 1 is produced by our Mammalian expression system and the target gene encoding Ala35-Thr794 is expressed with a 6His tag at the C-terminus.
: Accessories not included.
: Shipped Via FedEx
: Shipped at ambient temperature via FedEx envelope.
: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
- Protein/Peptide Type:Recombinant
- Protein/Peptide Name:Leucine-rich repeats and immunoglobulin-like domains protein 1
- Species:Human
Frequently Bought Together