Order Entry
Puerto Rico
ContactUsLinkComponent
Recombinant Mouse BMP Receptor IA, Bon Opus Biosciences
Recombinant Mouse BMP Receptor IA, Bon Opus Biosciences
Certificates
Recombinant Mouse BMP Receptor IA, Bon Opus Biosciences

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Mouse
  • Protein Synonyms:
    Bone morphogenetic protein receptor type-1A|Bmpr1a|BMP-4 receptor|Activin receptor-like kinase 3|BMPR-1A|SKR5|BMP-2|BMP type-1A receptor|Serine/threonine-protein kinase receptor R5|CD292|Acvrlk3
  • Protein/Peptide Name:
    ALK-3

Specifications

Frequently Bought Together