About this item
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
- Consistent Sizing options
- Datasheet Included
- High Purity, Low Endotoxin levels
Recombinant Human Annexin A8 is produced by our E.coli expression system and the target gene encoding Met1-Pro327 is expressed.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Enzyme type:Recombinant
- Species:Human
- Enzyme Name:Cyclin-Dependent Kinase 2