Vikan® Double Blade Ultra Hygiene Squeegee, 24", Remco Products
Supplier: Remco Products
Certificates
About this item
The double blade, non-porous rubber design of this squeegee is effective in removing water.
- Made with FDA compliant materials
- Autoclavable at 121 °C (250 °F)
The double blade, non-porous rubber design of this squeegee is effective in removing water from all types of floors; tiles, epoxy, or cement. It is very efficient in large food production facilities on walkways and expansive areas. The cartridge system blades can be removed for easy cleaning and replaced when worn out, making the model a very economical hygienic option.
Accessories information: Replacement squeegee blades available, see 76007-986, 76007-988, 76007-990, 76007-992, 76007-994, 76007-996, 76007-998, 76008-000, 76008-002.
Delivery: Dropshipping available.