Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-IgG F(c) Goat Polyclonal Antibody (DyLight® 650)
Anti-IgG F(c) Goat Polyclonal Antibody (DyLight® 650)
Catalog # 10149-920
Supplier:  Immunoreagents
undefined
Anti-IgG F(c) Goat Polyclonal Antibody (DyLight® 650)
Catalog # 10149-920
Supplier:  Immunoreagents
Supplier Number:  GTXHU-004-F2650NHSX

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG F(c)
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 650
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    1.0 mg
  • Epitope:
    F(c)
  • Cross Adsorption:
    Yes
  • Format:
    Purified IgG
  • Form:
    Lyophilized
  • Antibody Fragment:
    F(c)
  • Storage Buffer:
    10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
  • Storage Temperature:
    2-8 °C
  • Concentration:
    1.0 mg/ml (E 1% at 280 nm = 13.0)
  • Shipping Temperature:
    2-8 °C
  • Immunogen:
    Human IgG, Fc fragment
  • Tested Applications:
    Fluoresence ELISA, Fluorescence-Activated Cell Sorting
  • Purification:
    Affinity purified using solid phaseHuman IgG
  • Cat. No.:
    10149-920
  • Packaging:
    Glass vial
  • Supplier no.:
    IR1695

Specifications

About this item

  • Origin: US

Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human