Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-IgG Bovine Polyclonal Antibody
  10151-048
undefined
Anti-IgG Bovine Polyclonal Antibody
  10151-048
 :  BO-003-C

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Secondary
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Bovine
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western Blot:
    Yes
  • Size:
    10.0 mg
  • Form:
    Clear, colorless liquid, 0.2 µm filtered
  • Storage Temperature:
    2-8 °C
  • Concentration:
    > 4.5 mg/ml (E 1% at 280 nm = 13.0)
  • Shipping Temperature:
    2-8 °C
  • Tested Applications:
    FACS, Immunodiffusion, Labeling
  • Purification:
    Protein A
  • Cat. No.:
    10151-048
  • Supplier no.:
    IR2381

 

 

  • Origin: US

Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Bovine
Isotype:
Reactivity: