Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Goat IgG Polyclonal Antibody
Anti-Goat IgG Polyclonal Antibody
Catalog # 10151-028
Supplier:  Immunoreagents
undefined
Anti-Goat IgG Polyclonal Antibody
Catalog # 10151-028
Supplier:  Immunoreagents
Supplier Number:  GT-003-N

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Secondary
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western Blot:
    Yes
  • Size:
    1.0 mg
  • Storage Temperature:
    2-8 °C
  • Shipping Temperature:
    2-8 °C
  • Tested Applications:
    FACS, Immunodiffusion, Labeling
  • Cat. No.:
    10151-028
  • Supplier no.:
    IR2371

Specifications

About this item

  • Origin: US

Sterile

Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: