ContactUsLinkComponent
Artificial CSF Perfusion Fluid, Harvard Apparatus
Artificial CSF Perfusion Fluid, Harvard Apparatus
Catalog # 103244-114
Supplier:  Harvard Apparatus
CAS Number:  
Artificial CSF Perfusion Fluid, Harvard Apparatus
Catalog # 103244-114
Supplier:  Harvard Apparatus
Supplier Number:  59-7316
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Artificial CSF perfusion fluid
  • Cat. No.:
    103244-114
Specifications
About this item
Artificial cerebrospinal fluid (aCSF) is commonly used when sampling from brain interstitial fluid. This solution closely matches the electrolyte concentrations of CSF.
  • Prepared from high purity water and analytical grade reagents
  • Microfiltered and sterile
Final ion concentrations (in mm): Na 150; K 3.0; Ca 1.4; Mg 0.8; P 1.0; Cl 155.
Packaging: Supplied as a package of six 25 ml vials.