Lysobacter Enzymogenes Endoproteinase LysC
Catalog # 103258-578
Supplier: New England Biolabs (NEB)
Small Business Enterprise
CAS Number:
Specifications
- Species:Lysobacter enzymogenes
- Size:20 µg
- Enzyme Name:Endoproteinase LysC
- Cat. No.:103258-578
- Supplier no.:P8109S
Specifications
About this item
Endoproteinase LysC is a serine endoproteinase, isolated from Lysobacter enzymogenes, that cleaves peptide bonds at the carboxyl side of lysine. LysC is a sequencing grade enzyme and is suitable for proteomics and glycobiology applications.
- Analyze complex proteomes
- Compatible for simultaneous co-digestion with Trypsin-ultra Mass Spectrometry Grade
- Suitable for both in-gel and solution digests
- Supplied lyophilized
- Optimal activity and stability for up to 12 months
Delivery: Supplied freeze dried from Tris-HCl buffer, pH 8.