Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Support Stand, Lightnin

Support Stand, Lightnin

Support stand, 610 mm (shaft), Laboratory mixers, ⁵/₈Ø×24"H, 16 mm (shaft)
Catalog # 58958-095
Supplier:   Lightnin
Supplier #:  829453
Support Stand, Lightnin
Catalog # 58958-095
Supplier:  Lightnin
Support Stand, Lightnin

Support stand, 610 mm (shaft), Laboratory mixers, ⁵/₈Ø×24"H, 16 mm (shaft)

Price
$888.29
Log in for availability
Each
Product Details & Documents

This high density ring stand is mainly designed for use with laboratory mixers.

Specifications
  • Ø:
    16 mm (shaft)
  • Description:
    Support stand
  • Dimensions:
    ⁵/₈Ø×24"H
  • Length:
    610 mm (shaft)
  • For use with:
    Laboratory mixers