Order Entry
Puerto Rico
ContactUsLinkComponent
TSKgel® Protein A-5PW HPLC Columns, Tosoh Biosciences
TSKgel® Protein A-5PW HPLC Columns, Tosoh Biosciences
Catalog # 103257-544
Supplier:  Tosoh Bioscience
TSKgel® Protein A-5PW HPLC Columns, Tosoh Biosciences
Catalog # 103257-544
Supplier:  Tosoh Bioscience
Supplier Number:  23483

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Column Format:
    Analytical
  • Ø int.:
    4.60 mm
  • Particle Size:
    20 µm
  • Pore size:
    1000 Å
  • Description:
    TSKgel Protein A-5PW
  • Length:
    35 mm
  • Column hardware:
    PEEK
  • Cat. No.:
    103257-544
  • Packing material:
    Hydroxylated Methacrylic Polymer Beads
  • Exclusion limit:
    1000 kDa
  • Supplier no.:
    23483

Specifications

About this item

TSKgel® Protein A-5PW columns have been designed for the rapid separation and robust quantification of a variety of antibodies. Monoclonal antibodies from harvested cell culture media can be captured and accurately quantitated in less than 2 minutes per injection. The column can be used for more than 2000 injections without regeneration or cleaning.

  • Packed with hydroxylated methacrylic polymer beads

Designed with a high degree of crosslinking, which allows high flow rate for chromatography while still maintaining chromatographic efficiency, peak width and resolution. The recombinant protein A ligand is a code-modified hexamer of the C domain. An enhanced rProtein A ligand is bound to the TSKgel® 5PW base bead via multipoint attachment resulting in excellent base stability in 0.1 mol/L NaOH.

The wide range loading capacity of the TSKgel® Protein A-5PW column can accurately determine the titer of mAb at various stages of cell culture media processing. The low level of protein A leaching makes this column a good candidate for small scale purification of mAbs for initial characterization.