ContactUsLinkComponent
Scoop/Ladle, HDPE, Dynalon
  76210-886
 :  Dynalon
undefined
Scoop/Ladle, HDPE, Dynalon
  76210-886
 :  Dynalon
 :  108085-50
  • Capacity:
    50 ml
  • Length:
    9" (Pole)
  • Material:
    HDPE
  • Cat. No.:
    76210-886
Plastic ladle dippers with a heavy-duty bowl for sampling and dispensing corrosive liquids.
  • Handy for sampling and dispensing corrosive liquids
Fabricated in the USA from high density polyethylene, these plastic ladle dippers feature a high-temperature, extra heavy-duty bowl with securely welded solid handle.