ContactUsLinkComponent
T2 / HT2 Immunoaffinity Column, Eagle Biosciences, Inc.
  103388-992
T2 / HT2 Immunoaffinity Column, Eagle Biosciences, Inc.
  103388-992
 :  BTTE316005
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    T2 / HT2 Immunoaffinity Column, 10 columns
  • Cat. No.:
    103388-992
The T2/HT2 determination in food and feed focuses on extract using immunoaffinity column (IAC).
  • Sensitivity: 0.2 to 400 ng
  • Dynamic range: 0.2 to 400 ng
  • Sample size: 10 columns
This instruction of T2/HT2 determination in food and feed focuses on the enrichment step of extract using immunoaffinity column (IAC) and quantification with HPLC. Accepted laboratory extraction methods could be maintained. Full performance of the IAC column is given if pronounced criteria regarding organic solvent tolerance, elution process of analyte and working range of column is followed. Many pretreatment methods for T2/HT2 in food and feed show low sensitivity because of interferring substances if problematic matrices are applied. This method of content determination of T2/HT2 combines the high selectivity of an immunoaffinity column (IAC) with its potential to concentrate elute and additional step of purification of labelled T2/HT2 by HPLC column. As said before, this instruction focuses on the handling with the IAC column. For the commodity extraction step a literature method is given. Please see below. The given apparatus (e.g. HPLC system) might serve as example among other possibilities