Orders LinkContactUsLinkComponent
Estradiol Immunoaffinity Column, Eagle Biosciences, Inc.
Estradiol Immunoaffinity Column, Eagle Biosciences, Inc.
Estradiol Immunoaffinity Column, 10 columns
Catalog # 103388-986
Marketsource
Supplier:   Eagle Biosciences
Supplier #:  BTES320005
Estradiol Immunoaffinity Column, Eagle Biosciences, Inc.
Catalog # 103388-986
Supplier:  Eagle Biosciences
Marketsource
Estradiol Immunoaffinity Column, Eagle Biosciences, Inc.
Estradiol Immunoaffinity Column, 10 columns
Price
$372.69
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
This method to detect Estrogens combines immunoaffinity columns with elute and purification by HPLC.
  • Sample Size: 10 columns
Many methods of Estrogens determination based on HPLC-UV detection show low selectivity if problematic matrices are applied. This method of content determination of Estrogens combines the high selectivity of immunoaffinity columns with its high potential to concentrate elute and of purification by HPLC column.
The following instruction gives a optimized protocol for enrichment of very low contents in the pg/g range. In this case a mass detector might be best suited. Please notice, that this instruction focuses on the handling with the IAC column. The given apparatus and detection (e.g. HPLC-FLD system) might serve as example among other possibilities.
Specifications
  • Description:
    Estradiol Immunoaffinity Column, 10 columns