ContactUsLinkComponent
Porphyrins HPLC Assay, Eagle Biosciences
  102994-892
 :  
Porphyrins HPLC Assay, Eagle Biosciences
  102994-892
 :  PPR34-H100
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Porphyrins HPLC Assay
  • Size:
    1 kit
  • Environmentally Preferable:
  • Cat. No.:
    102994-892
  • Supplier no.:
    PPR34-H100
The Porphyrins HPLC Assay Kit is intended to determine porphyrins in the urine.
  • Incubation Time: 5 minutes
  • Sample Size: 1 mL
  • Sample Type: Urine
Hemoglobin, myoglobin and the cytochromes contain the heme structure as active site. The porphyrins are precursors of the heme molecules. They play an important role in the oxygen metabolism. The heme synthesis is located mainly in the erythrocyte cells. Glycin and succinyl-CoA are condensed to δ-aminolävulinic acid. Two molecules of δ-aminolävulinic are then condensed to porphobilinogen. The porphyrin ring is built up by four porphobilinogen molecules.