ContactUsLinkComponent
Homocysteine HPLC Column, Eagle Biosciences, Inc.
Homocysteine HPLC Column, Eagle Biosciences, Inc.
Catalog # 102994-516
Supplier:  Eagle Biosciences
Homocysteine HPLC Column, Eagle Biosciences, Inc.
Catalog # 102994-516
Supplier:  Eagle Biosciences
Supplier Number:  IC2801RP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Homocysteine HPLC Column, 1 column
  • Cat. No.:
    102994-516
  • Supplier no.:
    IC2801RP
Specifications
About this item
Homocysteine HPLC column can be used with the HPLC assay kit and can be used for 500 to 1000 samples.
  • For use with homocysteine HPLC assay kit
Homocysteine is an amino acid, which is produced out of methionine. It is not ingested by nutrition. Homocysteine is rapidly degraded to cysteine when vitamin B6, folic acid and vitamin B12 are sufficiently available. Increased levels of homocysteine will damage heart and blood vessels. Meanwhile it is an established risk marker for cardiovascular disease. US studies demonstrate that 40% of heart attacks are caused by increased homocysteine levels. Also the risk for stroke is increased by elevated homocysteine concentrations in plasma. A participation of homocysteine in Alzheimer and Parkinson diseases are under discussion.