ContactUsLinkComponent
Vitamin K1 HPLC Column, Eagle Biosciences, Inc.
Vitamin K1 HPLC Column, Eagle Biosciences, Inc.
Catalog # 102994-508
Supplier:  Eagle Biosciences
Vitamin K1 HPLC Column, Eagle Biosciences, Inc.
Catalog # 102994-508
Supplier:  Eagle Biosciences
Supplier Number:  IC2400RP
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Vitamin K1 HPLC Column, 1 column
  • Cat. No.:
    102994-508
  • Supplier no.:
    IC2400RP
Specifications
About this item
Vitamin K1 HPLC assay kit column can be used with the HPLC kit and have about 500 to 1000 samples.
  • For use with Vitamin K1 HPLC assay kit
Vitamin K works as cofactor of an oxygen dependent carboxylase, which carboxylates specific glutamic acid residues in g-position. Because of that modification, blood clotting factors can be activated by cleavage. In bone metabolism the g-carboxylation enables osteocalcin to bind to hydroxyapatite of the bone matrix. Vitamin K can be stored only for a short time, whereby deficiency occur after a view days. A lack of vitamin K mainly manifests in clotting disorders. Beside this, disorders in bone metabolism caused by a deficient modification of osteocalcin have been reported. The Eagle Biosciences Vitamin K1 HPLC Assay kit makes it possible to determine the vitamin in an easy, fast and precise method.
Ordering information: The kit includes all reagents for the preparation and separation of the samples with exception of the columns (IC2400rp) and the controls (IC2400ko).