- Description:Vitamin B2 HPLC Column, 1 column
- Cat. No.:102994-504
- Supplier no.:IC2300RP
Vitamin B2 HPLC assay kit column can be used for about 500 to 1000 samples.
- For use with vitamin B2 HPLC assay kit
Vitamin B2 is a collective term for the molecules Riboflavin, Flavin Adenine Dinucleotide (FAD) and Flavin Adenine Mononucleotide (FMN). Riboflavin belongs to the group of Flavonoids (plant pigments). It only can be synthesized by plants and microorganisms. Humans and animals ingest it via the gut and convert it to the coenzymes FAD and FMN. They are distributed within the whole body with maximum concentrations in liver, kidney and heart.