ContactUsLinkComponent
Vitamin B1 (Thiamin) HPLC Column, Eagle Biosciences, Inc.
Vitamin B1 (Thiamin) HPLC Column, Eagle Biosciences, Inc.
Catalog # 102994-500
Supplier:  Eagle Biosciences
Vitamin B1 (Thiamin) HPLC Column, Eagle Biosciences, Inc.
Catalog # 102994-500
Supplier:  Eagle Biosciences
Supplier Number:  IC2201RP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Vitamin B1 (Thiamin) HPLC Column, 1 column
  • Cat. No.:
    102994-500
  • Supplier no.:
    IC2201RP
Specifications
About this item
Vitamin B1 HPLC assay kit column can be used for about 500 to 1000 samples.
  • For use with Vitamin B1 (Thiamin) HPLC assay kit
The Eagle Biosciences Vitamin B1 (Thiamin) HPLC assay makes it possible to determine the vitamin in an easy, fast and precise method. The kit includes all reagents in ready to use form for preparation and separation of the samples with exception of the columns (IC2201rp) and the controls (IC2201ko). Thiamin is found free and as mono-, di- and triphosphate. The different forms can be transferred into each other. The metabolic active form is thiamin diphosphate in the body and thiamin triphosphate in the brain. Thiamin diphosphate (TDP) works as thermostable coenzyme in many enzymes of the carbohydrate and amino acid metabolism. In most reactions TDP catalyzes the cleavage of C-C bonds. In the nervous system vitamin B1 is needed for the stimulation of nerve cells.