ContactUsLinkComponent
Vitamin B6 HPLC Column, Eagle Biosciences, Inc.
Vitamin B6 HPLC Column, Eagle Biosciences, Inc.
Catalog # 102994-496
Supplier:  Eagle Biosciences
Vitamin B6 HPLC Column, Eagle Biosciences, Inc.
Catalog # 102994-496
Supplier:  Eagle Biosciences
Supplier Number:  IC2100RP
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Vitamin B6 HPLC Column, 1 column
  • Cat. No.:
    102994-496
  • Supplier no.:
    IC2100RP
Specifications
About this item
Vitamin B6 HPLC assay kit column can be used for about 500 to 1000 samples.
  • For use with Vitamin B6 HPLC assay kit
The vitamins pyridoxin, pyridoxal and pyridoxamin and the appropriate phosphate products are summarized as vitamin B6. All forms can be transformed into the active form pyridoxal-5-phosphate. Vitamin B6 functions as a coenzyme and is essential for more than 50 reactions in the protein, carbohydrate and fatty acid metabolism thereby synthesizing, transforming or degrading amino acids. In protein metabolism vitamin B6 supports the resorption of amino acids and their transport into the cells. Furthermore vitamin B6 contributes to the synthesis of neurotransmitters and amine products (histamine).