ContactUsLinkComponent
Vitamin A & Vitamin E HPLC Column, Eagle Biosciences, Inc.
Vitamin A & Vitamin E HPLC Column, Eagle Biosciences, Inc.
Catalog # 102994-480
Supplier:  Eagle Biosciences
Vitamin A & Vitamin E HPLC Column, Eagle Biosciences, Inc.
Catalog # 102994-480
Supplier:  Eagle Biosciences
Supplier Number:  IC1600RP
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Vitamin A & Vitamin E HPLC Column, 1 column
  • Cat. No.:
    102994-480
  • Supplier no.:
    IC1600RP
Specifications
About this item
The Vitamin A & Vitamin E HPLC column is can be utilized for approximately 500 to 1000 samples.
  • For use with Vitamin A and E HPLC assay kit
Vitamin A and E are fat soluble vitamins. Both are stored in the body, thus overdosing is possible. Vitamin A plays an important role in the visual process. It is mainly taken up as pre vitamin A by nutrition. In the liver it is processed to vitamin A. Retinal is involved in the visual process as prosthetic group of the rhodopsin molecule. Beside this, vitamin A (Retinol) is important for the growth of children and the growth of skin and mucous membranes. As an antioxidant, retinol protects the body against free radicals and reactive oxygen species. Overdosing leads to headache, vertigo, sickness and vomit. Disturbance of the nervous system, skin disease and loss of hair have also been reported.