ContactUsLinkComponent
Biotin Immunoaffinity Column, Eagle Biosciences, Inc.
Biotin Immunoaffinity Column, Eagle Biosciences, Inc.
Catalog # 102994-022
Supplier:  Eagle Biosciences
Biotin Immunoaffinity Column, Eagle Biosciences, Inc.
Catalog # 102994-022
Supplier:  Eagle Biosciences
Supplier Number:  BTSA317005
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Biotin Immunoaffinity Column, 10 columns
  • Cat. No.:
    102994-022
  • Supplier no.:
    BTSA317005
Specifications
About this item
The Biotin immunoaffinity column detects Biotin in food products, cell culture samples by HPLC.
  • Sensitivity: 0.01 μg
  • Dynamic range: 0.01 to 5 μg
Fast and accurate content determination of biotin in vitamin tablets, liquid vitamin preparations, cell culture extracts, and more by combination of affinity chromatography, HPLC-FLD and post column derivatisation with fluorescein-streptavidin conjugate as fluorophor.