ContactUsLinkComponent
Ochratoxin Immunoaffinity Column, Eagle Biosciences, Inc.
Ochratoxin Immunoaffinity Column, Eagle Biosciences, Inc.
Catalog # 102994-020
Supplier:  Eagle Biosciences
Ochratoxin Immunoaffinity Column, Eagle Biosciences, Inc.
Catalog # 102994-020
Supplier:  Eagle Biosciences
Supplier Number:  BTOT312005
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Ochratoxin Immunoaffinity Column, 10 columns
  • Cat. No.:
    102994-020
  • Supplier no.:
    BTOT312005
Specifications
About this item
Ochratoxin Immunoaffinity column focuses on extract using IAC and with quantification with HPLC.
  • Sensitivity: 0.02 to 2 ng OTA
  • Dynamic range: 0.02 to 2 ng
  • Sample size: 10 columns
This instruction of Ochratoxin determination in food and feed focuses on the enrichment step of extract using immunoaffinity column (Immunoaffinity Chromotography (IAC)) in combination with quantification with HPLC (High Performance Liquid Chromotography). Accepted laboratory extraction methods could be maintained. If the criteria of organic solvent tolerance, elution process of analyte, and working range of column are followed you will see full performance with the IAC column. If problematic matrices are applied, many pretreatment methods of Ochratoxin determination in food and feed will show low sensitivity because of interfering substances.
This method of content determination of ochratoxins combines the high selectivity of an immunoaffinity column (IAC) with its potential to concentrate eluate with the additional step of purification by HPLC column. Please notice that this instruction focuses on the handling with the IAC column. For the commodity extraction step please note that a literature method is provided. The given apparatus (e.g. HPLC system) might serve as example among other possibilities.