ContactUsLinkComponent
Aflatoxin Immunoaffinity Column, Eagle Biosciences, Inc.
Aflatoxin Immunoaffinity Column, Eagle Biosciences, Inc.
Catalog # 102994-014
Supplier:  Eagle Biosciences
Aflatoxin Immunoaffinity Column, Eagle Biosciences, Inc.
Catalog # 102994-014
Supplier:  Eagle Biosciences
Supplier Number:  BTAF310005
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Aflatoxin Immunoaffinity Column, 10 columns
  • Cat. No.:
    102994-014
  • Supplier no.:
    BTAF310005
Specifications
About this item
Aflatoxin Immunoaffinity column focuses on extract using IAC and quantification with HPLC.
  • Sensitivity: 0.02 to 3 ng AFL
  • Dynamic range: 0.02 to 3 ng AFL
  • Sample size: 10 columns
This instruction of Aflatoxin total determination in food and feed focuses on the enrichment step of extract
using immunoaffinity column (Immunoaffinity Chromotography (IAC)) in combination with quantification with
HPLC (High Performance Liquid Chromotography). Accepted laboratory extraction methods should be followed throughout the procedure. If the criteria of organic solvent tolerance, elution process of analyte, and working range of column are followed you will see full performance with the IAC column.
If problematic matrices are applied, many pretreatment methods of aflatoxin (AFG2, AFG1, AFB2 and AFB1)
determination in food and feed will show low sensitivity because of interfering substances. This method of
content determination of aflatoxins combines the high selectivity of an immunoaffinity column (IAC) with its
potential to concentrate eluate with the additional step of purification by HPLC column. Please notice that this instruction focuses on the handling with the IAC column. For the commodity extraction step please note that a literature method is provided. The given apparatus (e.g. HPLC system) might serve as example among other possibilities.