Orders LinkContactUsLinkComponent
Rig Lizard® 2030 Gloves, HexArmor
Rig Lizard® 2030 Gloves, HexArmor
Supplier:   HexArmor
Rig Lizard® 2030 Gloves, HexArmor
Supplier:  HexArmor
Certificates
Rig Lizard® 2030 Gloves, HexArmor
1 Options Available Below
Product Details & Documents
The Rig Lizard® 2030 delivers maximum grip and dexterity without sacrificing protection. It features a hi-vis silicone palm for superior grip and heat resistance, and the patented high-flex IR-X® Impact Exoskeleton™ protects the entire back- of-hand from smash and pinch injuries.
  • SuperFabric® brand material palm provides ANSI/ISEA cut level A6 resistance for some of the highest cut protection available in the industry (interior layer)
  • Glove was tested to the EN407 standard and received a level 3
  • Durable synthetic leather palm with heat-resistant silicone pattern provides superior grip and dexterity
  • Back-of-hand IR-X® Impact Exoskeleton™ with high-flex design keeps hands safe from smash and impact hazards
  • SlipFit® cuff with Velcro® closure for a secure and comfortable fit
  • Hi-vis color scheme increases visual awareness
  • Launderable
The Rig Lizard® 2030 with A6 cut resistance is a leader in the oil, gas, and mining industries. The durable TP-X® grip technology provides the ultimate grip in wet/dry applications while the patented full back-of-hand IR-X® Impact Exoskeleton™ keeps hands safe from smash and pinch hazards.
Specifications for Product Family
Specifications
  • Catalog No:
  • 76207-892
  • 76207-900
  • 76207-902
  • Ambidextrous/Hand-specific:
  • Hand-Specific
  • Disposable/Reusable:
  • Reusable
  • Latex/Latex-free:
  • Latex-Free
  • Powdered/Powder-free:
  • Powder-Free
  • EN Norm:
  • En388 4532
  • En388 4532
  • En388 4532
Specifications
Product Family Options
Product Information
  • Model
    Glove size
    Glove color
    Length
    Availability
    Price
  • Rig Lizard® 2030
    Small
    Natural
    10"
    Log in for availability
    Log in for availability
    Pair
    Pair - $104.88
    $104.88
    Pair
       $104.88
    Supplier Number: 2030X-S(7)
    Specifications:
    More
    Specifications
    Cat. No.
    76207-892
    Ambidextrous/Hand-specific
    Hand-Specific
    Disposable/Reusable
    Reusable
    Latex/Latex-free
    Latex-Free
    Powdered/Powder-free
    Powder-Free
    EN Norm
    En388 4532
  • Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    76207-900
    Ambidextrous/Hand-specific
    Disposable/Reusable
    Latex/Latex-free
    Powdered/Powder-free
    EN Norm
    En388 4532
  • Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    76207-902
    Ambidextrous/Hand-specific
    Disposable/Reusable
    Latex/Latex-free
    Powdered/Powder-free
    EN Norm
    En388 4532