ContactUsLinkComponent
Econo™ Metzenbaum-Nelson Bariatric Dissecting Scissors, Floor Grade, Sklar®
Econo™ Metzenbaum-Nelson Bariatric Dissecting Scissors, Floor Grade, Sklar®
Catalog # 76192-454
Supplier:  Sklar
undefined
Econo™ Metzenbaum-Nelson Bariatric Dissecting Scissors, Floor Grade, Sklar®
Catalog # 76192-454
Supplier:  Sklar
Supplier Number:  21-354
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    9"
  • Material:
    Stainless steel
  • Curvature:
    Curved
  • Cat. No.:
    76192-454
  • Blade type:
    Blunt, Smooth
Specifications
About this item
Sklar® Metzenbaum-Nelson Dissecting Scissors are most commonly used for dissecting and cutting tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar® Metzenbaum-Nelson Dissecting Scissors are not recommended for cutting sutures, drains, or heavy tissue. These scissors tend to be longer than standard Metzenbaum scissors and are ideal for use in thoracic procedures.