Specifications
- Length:5"
- Material:Stainless steel
- Curvature:Straight
- Cat. No.:76192-960
Specifications
About this item
The Sklar® Harvey Suture Wire Cutting Scissors are non-ratcheted, finger ring scissor used for cutting sutures in preparation for removal.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
Blades are much shorter than the shanks, and the jaws have one serrated saw edge. The angled parrot beak style, like the Sklar Edge™ TC wire cutting scissors, is more frequently used than the straight Harvey style.