ContactUsLinkComponent
Econo™ Simpson Uterine Sound, Floor Grade, Sklar®
Econo™ Simpson Uterine Sound, Floor Grade, Sklar®
Catalog # 76192-466
Supplier:  Sklar
undefined
Econo™ Simpson Uterine Sound, Floor Grade, Sklar®
Catalog # 76192-466
Supplier:  Sklar
Supplier Number:  21-382
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Length:
    12.5"
  • Material:
    Silver Plated
  • Environmentally Preferable:
  • Curvature:
    Curved
  • Cat. No.:
    76192-466
Specifications
About this item
This is a malleable, OB/GYN instrument used for probing and dilating the uterus, as well as measuring the length of the cervical canal.
  • Industry standard
Round and thin with a small nodule at the tip and a thumb grip handle. It is very similar to the Sims uterine sound.
Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.