- Length:11"
- Material:Stainless Steel with Tungsten Carbide
- Curvature:Straight
- Cat. No.:76192-674
The Sklar® Bihrle needle holder/clamp with tungsten carbide is a ratcheted finger ring instrument used in fixation and suturing during cancer related prostatetomies or cystectomies.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
The angled jaw eases exposure and visualization. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.