Specifications
- Length:14"
- Material:Stainless Steel with Tungsten Carbide
- Curvature:Straight
- Cat. No.:76192-592
Specifications
About this item
The Sklar® TC DeBakey Needle Holder is a ratcheted, finger-ring instrument used to hold and guide small to medium size needles and suture materials.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
The DeBakey Needle Holder has a gently tapered, blunt tip and is more delicate than the Mayo-Hegar, but slightly less delicate than Webster, Halsey, or Crile-Wood needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. Tungsten Carbide instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.