Specifications
- Length:4.25"
- Material:Stainless Steel with Tungsten Carbide
- Curvature:Straight
- Cat. No.:76192-390
Specifications
About this item
The Sklar® TC Martini needle holder is a smooth jawed instrument that is less damaging to the needle.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
The TC Martini needle holder is available individually, or as a component of the Sklar® TMJ surgery set # 98-1464. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.