- Length:7"
- Material:Stainless Steel with Tungsten Carbide
- Curvature:Straight
- Cat. No.:76192-150
Sklar Edge™ TC Stella scissors are used in plastic surgery procedures.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
Sklar Edge™ TC Supercut Sklarhone™ scissors provide effortless, precise cutting. These scissors feature one knife-like razor edge and one micro-serrated edge along with the strength and durability of tungsten carbide. They can be used to cut both delicate and heavy tissue with minimal tearing or pulling. Sklar permanently bonds tungsten carbide inserts to the blades, which maximizes control and precision. These scissors are also designed to stay sharp for a significantly longer period of time than standard scissors.